Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PPP1R8 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-3658R-CY3
10371-490
Anti-PPP1R8 Rabbit Polyclonal Antibody (Cy3®)
Inhibitor subunit of the major nuclear protein phosphatase 1 (PP1). It has RNA binding activity but does not cleave RNA and may target PP1 to RNA associated substrates. May also be involved in pre mRNA splicing. Binds DNA and might act as a transcriptional repressor. Seems to be required for cell proliferation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PPP1R8
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty