Anti-PLEC Rabbit Polyclonal Antibody

Supplier: Bioss

BS-3640R
10375-800EA 649.69 USD
10375-800
Anti-PLEC Rabbit Polyclonal Antibody
Antibodies
Plectin interlinks intermediate filaments with microtubules and microfilaments and anchors intermediate filaments to desmosomes or hemidesmosomes. Could also bind muscle proteins such as actin to membrane complexes in muscle. May be involved not only in the crosslinking and stabilization of cytoskeletal intermediate filaments network, but also in the regulation of their dynamics.

Recommended Dilutions: IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: PLEC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR