Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PLXA2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-3638R-A647
10375-784
Anti-PLXA2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Plexin A2 is a coreceptor for SEMA3A and is necessary for signaling by class 3 semaphorins and subsequent remodeling of the cytoskeleton. This coreceptor plays a role in axon guidance, invasive growth and cell migration. Class 3 semaphorins bind to a complex composed of a neuropilin and a plexin in which the plexin modulates the affinity of the complex for specific semaphorins, and its cytoplasmic domain is required for the activation of down-stream signaling events in the cytoplasm.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PLXA2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty