Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SIAH1/2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss

BS-3634R-FITC
10371-182
Anti-SIAH1/2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Siah2 is a E3 Ubiquitin ligase protein that mediates ubiquitination and subsequent proteasomal degradation of target proteins. E3 ubiquitin ligases accept ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfer the ubiquitin to targets. Siah2 triggers the ubiquitin-mediated degradation of many proteins, including transcription regulators (e.g. POU2AF1, PML, NCOR1), cell surface receptors (e.g. DCC), antiapoptotic proteins (e.g. BAG1), and proteins involved in synaptic vesicle function in neurons (e.g. SYP). Siah2 has some overlapping function with SIAH1 but is able to trigger the ubiquitin-mediated degradation of TRAF2, whereas SIAH1 can not.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SIAH1/2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty