Anti-HSD11B2 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-3618R-CY7
10371-066EA 811.1 USD
10371-066
Anti-HSD11B2 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Catalyzes the conversion of cortisol to the inactive metabolite cortisone. Modulates intracellular glucocorticoid levels, thus protecting the nonselective mineralocorticoid receptor from occupation by glucocorticoids.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HSD11B2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR