Anti-ESM1 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-3615R-CY5.5
10373-744EA 811.1 USD
10373-744
Anti-ESM1 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Involved in angiogenesis; promotes angiogenic sprouting. May have potent implications in lung endothelial cell-leukocyte interactions.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ESM1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR