Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SSEA4 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-3609R-A350
10370-938
Anti-SSEA4 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
SSEA-4 (Stage-specific embryonic antigen 4) is a glycoprotein expressed early in embryonic development and in pluripotent stem cells. SSEA-4 can be used as a marker of Human Embryonic Stem Cells, Human Embryonic Carcinoma Cells and Human Embryonic Germ Cells. Monoclonal antibodies to this target have been widely used in the characterization of pluripotent stem cells. Mouse pluripotent stem cells are not recognised by anti-SSEA-4 antibodies but do express the antigen upon differentiation. 

Recommended Dilutions: Flow Cytometry: 1:20-100

Type: Primary
Antigen: SSEA4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty