Anti-MTF1 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-3601R-CY7
10370-850EA 776.17 USD
10370-850
Anti-MTF1 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Activates the metallothionein I promoter. Binds to the metal responsive element (MRE).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MTF1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR