Anti-MTF1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-3601R-A488
10370-836EA 775.93 USD
10370-836
Anti-MTF1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Activates the metallothionein I promoter. Binds to the metal responsive element (MRE).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MTF1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR