Anti-CXCR5 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss
BS-3598R-A555
10373-652EA 746.2 USD
10373-652
Anti-CXCR5 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Cytokine receptor that binds to B-lymphocyte chemoattractant (BLC). Involved in B-cell migration into B-cell follicles of spleen and Peyer patches but not into those of mesenteric or peripheral lymph nodes. May have a regulatory function in Burkitt lymphoma (BL) lymphomagenesis and/or B-cell differentiation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CXCR5
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR