Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GRIM19 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-3594R-CY5.5
10375-562
Anti-GRIM19 Rabbit Polyclonal Antibody (Cy5.5®)
A novel gene, Genes associated with Retinoid IFN induced Mortality (GRIM) GRIM19 gene was identified. Antisense expression of GRIM19 confers a strong resistance against IFN/RA induced death by reducing the intracellular levels of GRIM19 protein. Overexpression of GRIM19 enhances cell death in response to IFN/RA. GRIM19 is primarily a nuclear protein whose expression is induced by the IFN/RA combination. These data indicate that GRIM19 is a novel cell death regulatory molecule.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GRIM19
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty