Anti-GRIM19 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-3594R-CY3
10375-558EA 776.17 USD
10375-558
Anti-GRIM19 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
A novel gene, Genes associated with Retinoid IFN induced Mortality (GRIM) GRIM19 gene was identified. Antisense expression of GRIM19 confers a strong resistance against IFN/RA induced death by reducing the intracellular levels of GRIM19 protein. Overexpression of GRIM19 enhances cell death in response to IFN/RA. GRIM19 is primarily a nuclear protein whose expression is induced by the IFN/RA combination. These data indicate that GRIM19 is a novel cell death regulatory molecule.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GRIM19
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR