Anti-TNK1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss
BS-3587R-HRP
10373-642EA 746.42 USD
10373-642
Anti-TNK1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Tnk1 is a nonreceptor protein tyrosine kinase (NRPTK) and a new member of the Ack family of NRPTKs. Tnk1 is a ubiquitously expressed 47-kDa protein with autotyrosine kinase activity that is developmentally regulated during embryogenesis. Tnk1 is also upregulated following IL3 withdrawal from factor-dependent murine NSF/N1.H7 cells that undergo apoptosis, suggesting a role in growth inhibition. Data support a negative regulatory role for Tnk1 in regulating the Ras-Raf1-MAPK growth pathway by a mechanism that requires its autotyrosine kinase activity.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: TNK1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR