Anti-TCTP Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-3578R-A488
10370-560EA 811.1 USD
10370-560
Anti-TCTP Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
TCTP is involved in calcium binding and microtubule stabilization. It is an immunoglobulin E dependent histamine-releasing factor produced by lymphocytes of atopic children and present in biologic fluids of allergic patients. Both human and mouse recombinant HRF proteins caused histamine release from human basophils of a subpopulation of donors (referenced from OMIM).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TCTP
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR