Anti-SLC11A2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-3577R-A555
10375-416EA 811.1 USD
10375-416
Anti-SLC11A2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Important in metal transport, in particular iron. Can also transport manganese, cobalt, cadmium, nickel, vanadium and lead. Involved in apical iron uptake into duodenal enterocytes. Involved in iron transport from acidified endosomes into the cytoplasm of erythroid precursor cells. May play an important role in hepatic iron accumulation and tissue iron distribution. May serve to import iron into the mitochondria.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLC11A2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR