Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SSEA3 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-3575R-A647
10375-394
Anti-SSEA3 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
SSEA3 is a globoseries carbohydrate antigen present on both cell surface glycolipids and glycopeptides. It is found on the surface of mouse oocytes, becoming restricted to the intracellular mass of the early blastocyst and then to the primitive endoderm. It is present on the surface of human teratocarcinoma cells (EC), embryonic germ cells (EG) and embryonic stem cells (ES) but not mouse undifferentiated mouse cells of these types. SSEA3 expression decreases as human EC, ES and EG cells differentiate but increases on differentiation of mouse EC, ES and EG cells. It is also found on the surface of mouse erythrocytes.

Recommended Dilutions: IF(IHC-P): 1:50-200; IF(ICC)(1:50-200

Type: Primary
Antigen: SSEA3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty