Anti-SSEA3 Rabbit Polyclonal Antibody

Supplier: Bioss
BS-3575R
10375-386EA 505.24 USD
10375-386
Anti-SSEA3 Rabbit Polyclonal Antibody
SSEA3 is a globoseries carbohydrate antigen present on both cell surface glycolipids and glycopeptides. It is found on the surface of mouse oocytes, becoming restricted to the intracellular mass of the early blastocyst and then to the primitive endoderm. It is present on the surface of human teratocarcinoma cells (EC), embryonic germ cells (EG) and embryonic stem cells (ES) but not mouse undifferentiated mouse cells of these types. SSEA3 expression decreases as human EC, ES and EG cells differentiate but increases on differentiation of mouse EC, ES and EG cells. It is also found on the surface of mouse erythrocytes.

Recommended Dilutions: IHC-P: 1:100-500; IF(IHC-P): 1:50-200; IF(ICC)(1:50-200

Type: Primary
Antigen: SSEA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR