Anti-ADAMTS5 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-3573R-A350
10375-364EA 776.17 USD
10375-364
Anti-ADAMTS5 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Cleaves aggrecan, a cartilage proteoglycan, and may be involved in its turnover. May play an important role in the destruction of aggrecan in arthritic diseases. May play a role in proteolytic processing mostly during the peri-implantation period.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ADAMTS5
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR