Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RELB Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-3562R-CY5
10370-410
Anti-RELB Rabbit Polyclonal Antibody (Cy5®)
The NFKB complex consists of NFKB1 or NFKB2 bound to REL, RELA, or RELB. The NFKB complex is inhibited by I kappa B proteins (NFKBIA, or NFKBIB), which inactivate NF kappa B by trapping it in the cytoplasm. Phosphorylation of serine residues on the I kappa B proteins by kinases (IKBKA, or IKBKB,) marks them for destruction via the ubiquitination pathway, thereby allowing activation of the NF kappa B complex. For some genes, activation requires NFKB interaction with other transcription factors, such as STAT, AP1 (JUN), and NFAT.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RELB
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty