Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RPS6KA2 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-3553R-CY7
10375-162
Anti-RPS6KA2 Rabbit Polyclonal Antibody (Cy7®)
RSK3(Ribosomal S6 kinase 3)is a member of a family of 90-kDa ribosomal protein S6 kinases, which includes Rsk1, Rsk2 and Rsk3. These are broadly expressed serine/threonine protein kinases that are activated in response to mitogenic stimuli, including extracellular signal-regulated protein kinases Erk1 and Erk2. Rsk3 is a distinct isoform of p90Rsk that translocates to the cell nucleus, phosphorylates potential nuclear targets and may have a unique upstream activator. Several sites, such as Ser380, Thr359, Ser363 and Thr573, are important for its activation. Active Rsks appear to play a major role in transcriptional regulation by translocating to the nucleus and phosphorylating c Fos and CREB.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RPS6KA2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty