Anti-KSR1 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-3549R-CY3
10375-116EA 811.1 USD
10375-116
Anti-KSR1 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Location-regulated scaffolding protein connecting MEK to RAF. Promotes MEK and RAF phosphorylation and activity through assembly of an activated signaling complex. By itself, it has no demonstrated kinase activity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KSR1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR