Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DTNB Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-3548R-A647
10370-322
Anti-DTNB Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Dystrobrevin beta is a component of the dystrophin-associated protein complex and interacts with dystrophin short form DP71, syntrophins SNTG1 and SNTG2. It localizes to the sarcolemma and its disruption is associated with various forms of muscular dystrophy. 4 isoforms produced by alternative splicing have been identified.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DTNB
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty