Anti-PNK1/PNKP Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-3542R-CY5.5
10375-032EA 811.1 USD
10375-032
Anti-PNK1/PNKP Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Mammalian PNK catalyzes the phosphorylation of DNA at 5'-hydroxyl termini and can dephosphorylate its 3'-phosphate termini. It plays an important function in DNA repair following ionizing radiation or oxidative damage.PNK has been reported to participate in the repair of DNA-double strand breaks via PARP-1-dependent nonhomologous end-joining.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PNK1/PNKP
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR