Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PNK1/PNKP Rabbit Polyclonal Antibody (Biotin)

Supplier: Bioss

BS-3542R-BIO
10373-556
Anti-PNK1/PNKP Rabbit Polyclonal Antibody (Biotin)
Mammalian PNK catalyzes the phosphorylation of DNA at 5'-hydroxyl termini and can dephosphorylate its 3'-phosphate termini. It plays an important function in DNA repair following ionizing radiation or oxidative damage.PNK has been reported to participate in the repair of DNA-double strand breaks via PARP-1-dependent nonhomologous end-joining.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: PNK1/PNKP
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty