Anti-PXN Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-3539R-CY5.5
10375-008EA 811.1 USD
10375-008
Anti-PXN Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Paxillin is a 64 kDa cytoskeletal adapter protein involved in organisation and function of focal adhesions, which are critical to cell adhesion and migration. This in turn plays a role in a wide variety of processes including embryogenesis, organogenesis, wound repair, inflammation and cancer. Paxillin contains LD motifs, LIM domains, SH3 and SH2 binding domains that serve as docking sites for cytoskeletal proteins, tyrosine kinases (e.g., FAK, Pyk 2, Src), serine/threonine kinases, GTPase activating proteins and other adaptor proteins (e.g., Actin, Vinculin, Crk).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PXN
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR