Anti-PLK1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-3535R-A555
10374-902EA 811.1 USD
10374-902
Anti-PLK1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
PLK1 (polo-like kinase 1) is a member of the serine/threonine protein kinase family, cdc5/polo subfamily. PLK1 contains two polo box domains with a predicted molecular weight of 68 kDa. PLK1 has been shown to regulate cdc2/cyclin B through phosphorylation and activation of cdc25c phosphatase. PLK1 is modified by phosphorylation at Threonine 210. PLK1 may also be required for cell division. Depletion of PLK1 results in apoptosis and deregulation of expression of PKL1 is correlated with development of many malignancies.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PLK1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR