Anti-PFK2 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-3528R-CY3
10370-182EA 776.17 USD
10370-182
Anti-PFK2 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Synthesis and degradation of fructose 2,6-bisphosphate.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PFK2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR