Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PEA15 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-3524R-A555
10370-134
Anti-PEA15 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
PED/PEA 15 (Phosphoprotein Enriched in Diabetes/Phosphoprotein Enriched in Astrocytes 15 kDa) is a widely expressed 15 kDa protein comprised of an N terminal region containing a canonical Death Effector Domain (DED) sequence and a nuclear export signal, and a C terminal region containing two serine phosphorylation sites. PED/PEA 15 has been implicated in the regulation of multiple cellular processes including apoptosis, integrin activation, and insulin sensitive glucose transport in insulin responsive cells. Phosphorylation of both serine 104 (a Protein Kinase C site) and serine 116 (a substrate of CaMKII and Akt) is required for PED/PEA 15 function.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PEA15
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty