Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PAK4 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-3517R-A555
10370-078
Anti-PAK4 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
p21-activated kinases (PAKs) belong to the family of serine/threonine kinases involved in the control of various cellular processes, including the cell cycle, dynamics of the cytoskeleton, apoptosis, oncogenic transformation, and transcription. All PAK family members are characterized by the presence of p21-binding domain. p21-activated kinases are regulated by the small GTP-binding proteins Rac and Cdc42, and lipids, which stimulate autophosphorylation and phosphorylation of exogenous substrates. Serine (Ser-474) is the likely autophosphorylation site in the kinase domain of PAK4 in vivo. Phosphospecific directed against serine 474 detect activated PAK4 on the Golgi membrane when PAK4 is co-expressed with activated Cdc42. Current data strongly implicates PAK-4 in oncogenesis. PAK4 is frequently overexpressed in human tumor cell lines of various tissue origins.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PAK4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty