Anti-FCGR1A Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-3511R-CY5.5
10370-040EA 811.1 USD
10370-040
Anti-FCGR1A Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
High affinity receptor for the Fc region of immunoglobulins gamma. Functions in both innate and adaptive immune responses.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CD64/IGFR1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR