Anti-GRIN2A Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss
BS-3507R-A488
10374-496EA 746.42 USD
10374-496
Anti-GRIN2A Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
NMDA receptor subtype of glutamate-gated ion channels possesses high calcium permeability and voltage-dependent sensitivity to magnesium. Activation requires binding of agonist to both types of subunits.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NMDAR2A
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR