Anti-MYT1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-3505R-A350
10373-488EA 811.1 USD
10373-488
Anti-MYT1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Myt1 is a zinc finger protein that is known to interact with the co-repressor Sin3B and also HDAC1 and HDAC2. The Myt1 family, including Myt1 and Myt1L, exemplifies a class of neural sequence specific transcription factors that actively recruit HDACs to selected genes during CNS development.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MYT1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR