Anti-SYN2 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-3502R-CY5
10374-450EA 811.1 USD
10374-450
Anti-SYN2 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Synapsin II is a member of the synapsin gene family. Synapsins are neuronal phosphoproteins which associate with the cytoplasmic surface of synaptic vesicles. Family members are characterized by common protein domains, and they are implicated in synaptogenesis and the modulation of neurotransmitter release, suggesting a potential role in several neuropsychiatric diseases. This member of the synapsin family is a neuron-specific phosphoprotein that selectively binds to small synaptic vesicles in the presynaptic nerve terminal.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SYN2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR