Anti-SYN1 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-3501R-CY5
10374-426EA 776.17 USD
10374-426
Anti-SYN1 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Neuronal phosphoprotein that coats synaptic vesicles, binds to the cytoskeleton, and is believed to function in the regulation of neurotransmitter release. The complex formed with NOS1 and CAPON proteins is necessary for specific nitric-oxid functions at a presynaptic level.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SYN1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR