Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CDC25C Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-3482R-CY5
10374-306
Anti-CDC25C Rabbit Polyclonal Antibody (Cy5®)
Functions as a dosage-dependent inducer in mitotic control. Tyrosine protein phosphatase required for progression of the cell cycle. When phosphorylated, highly effective in activating G2 cells into prophase. Directly dephosphorylates CDK1 and activates its kinase activity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CDC25C
Clonality: Polyclonal
Clone: 0
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty