Anti-CDC25C Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-3482R-A350
10372-846EA 811.1 USD
10372-846
Anti-CDC25C Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Functions as a dosage-dependent inducer in mitotic control. Tyrosine protein phosphatase required for progression of the cell cycle. When phosphorylated, highly effective in activating G2 cells into prophase. Directly dephosphorylates CDK1 and activates its kinase activity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CDC25C
Clonality: Polyclonal
Clone: 0
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR