Anti-YAP1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss

BS-3476R-FITC
10373-370EA 776.19 USD
10373-370
Anti-YAP1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
YAP1 (also known as Yes associated protein 1) was originally identified as a transcription factor that binds to the SH3 domain of the YES kinase (a Src protein kinase). More recently it has been identified as a candidate oncogene that promotes tumorigenesis in many different types of cancer.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: YAP1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR