Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-YAP1 Rabbit Polyclonal Antibody (Biotin)

Supplier: Bioss

BS-3476R-BIO
10369-608
Anti-YAP1 Rabbit Polyclonal Antibody (Biotin)
YAP1 (also known as Yes associated protein 1) was originally identified as a transcription factor that binds to the SH3 domain of the YES kinase (a Src protein kinase). More recently it has been identified as a candidate oncogene that promotes tumorigenesis in many different types of cancer.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: YAP1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty