Anti-YAP1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-3476R-A647
10369-606EA 811.12 USD
10369-606
Anti-YAP1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
YAP1 (also known as Yes associated protein 1) was originally identified as a transcription factor that binds to the SH3 domain of the YES kinase (a Src protein kinase). More recently it has been identified as a candidate oncogene that promotes tumorigenesis in many different types of cancer.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: YAP1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR