Anti-TH Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-3463R-A555
10373-260EA 811.12 USD
10373-260
Anti-TH Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Neuronal MarkerTyrosine hydroxylase is involved in the conversion of tyrosine to dopamine. As the rate-limiting enzyme in the synthesis of catecholamines, tyrosine hydroxylase has a key role in the physiology of adrenergic neurons. Tyrosine hydroxylase is regularly used as a marker for dopaminergic neurons, which is particularly relevant for research into Parkinson's disease.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TH
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR