Anti-Troponin I Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-3458R-CY5.5
10372-716EA 811.12 USD
10372-716
Anti-Troponin I Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Troponin T is the tropomyosin-binding subunit of troponin, the thin filament regulatory complex which confers calcium-sensitivity to striated muscle actomyosin ATPase activity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Troponin I
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR