Anti-Tuberin/TSC2 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-3442R-CY3
10369-162EA 811.12 USD
10369-162
Anti-Tuberin/TSC2 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Tuberin, or TSC2 (Tuberous sclerosis complex), is implicated as a tumor suppressor. It may function in vesicular transport, and may also play a role in the regulation of cell growth arrest and in the regulation of transcription mediated by steroid receptors. Interaction between hamartin (TSC1) and tuberin may facilitate vesicular docking. It specifically stimulates the intrinsic GTPase activity of the Ras related protein RAP1A and RAB5, suggesting a possible mechanism for its role in regulating cellular growth. Mutations in tuberin lead to constitutive activation of RAP1A in tumors. At least three isoforms of Tuberin exist.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Tuberin/TSC2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR