Anti-SYK Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-3434R-A647
10369-054EA 776.19 USD
10369-054
Anti-SYK Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
This gene encodes a member of the family of non-receptor type Tyr protein kinases. This protein is widely expressed in hematopoietic cells and is involved in coupling activated immunoreceptors to downstream signaling events that mediate diverse cellular responses, including proliferation, differentiation, and phagocytosis. It is thought to be a modulator of epithelial cell growth and a potential tumour suppressor in human breast carcinomas. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Mar 2010]

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SYK
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR