Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SYK Rabbit Polyclonal Antibody

Supplier: Bioss

BS-3433R
10372-604
Anti-SYK Rabbit Polyclonal Antibody
Syk (72 kDa) is a non receptor protein tyrosine kinase that plays an important role in immune receptor signal transduction and is implicated in endothelial cell functions, including cell growth and migration. SYK is a positive effector of BCR stimulated responses. It couples the B cell antigen receptor (BCR) to the mobilization of calcium ions either through a phosphoinositide 3 kinase dependent pathway, when not phosphorylated on tyrosines of the linker region, or through a phospholipase C gamma dependent pathway, when phosphorylated on Tyr 342 and Tyr 346. Thus the differential phosphorylation of SYK can determine the pathway by which BCR is coupled to the regulation of intracellular calcium ions. Alternate Names: p72syk; Spleen tyrosine kinase; Tyrosine protein kinase SYK.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: SYK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty