Anti-STAT4 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-3430R-CY3
10368-994EA 811.12 USD
10368-994
Anti-STAT4 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
The protein encoded by this gene is a member of the STAT family of transcription factors. In response to cytokines and growth factors, STAT family members are phosphorylated by the receptor associated kinases, and then form homo- or heterodimers that translocate to the cell nucleus where they act as transcription activators. This protein is essential for mediating responses to IL12 in lymphocytes, and regulating the differentiation of T helper cells. Mutations in this gene may be associated with systemic lupus erythematosus and rheumatoid arthritis. Alternate splicing results in multiple transcript variants that encode the same protein. [provided by RefSeq, Aug 2011].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: STAT4
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR