Anti-PTPN11 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-3405R-A488
10368-626EA 776.19 USD
10368-626
Anti-PTPN11 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Acts downstream of various receptor and cytoplasmic protein tyrosine kinases to participate in the signal transduction from the cell surface to the nucleus. Dephosphorylates ROCK2 at Tyr-722 resulting in stimulatation of its RhoA binding activity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PTPN11
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR