Anti-SGK1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-3395R-A488
10368-490EA 776.19 USD
10368-490
Anti-SGK1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
This gene encodes a serine/threonine protein kinase that plays an important role in cellular stress response. This kinase activates certain potassium, sodium, and chloride channels, suggesting an involvement in the regulation of processes such as cell survival, neuronal excitability, and renal sodium excretion. High levels of expression of this gene may contribute to conditions such as hypertension and diabetic nephropathy. Several alternatively spliced transcript variants encoding different isoforms have been noted for this gene. [provided by RefSeq, Jan 2009]

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SGK1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR