Anti-RPS6 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss

BS-3388R-HRP
10372-466EA 776.19 USD
10372-466
Anti-RPS6 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
May play an important role in controlling cell growth and proliferation through the selective translation of particular classes of mRNA.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: RPS6
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR