Anti-RPS6 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-3388R-A350
10368-368EA 776.19 USD
10368-368
Anti-RPS6 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
May play an important role in controlling cell growth and proliferation through the selective translation of particular classes of mRNA.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RPS6
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR