Anti-PLCB3 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-3341R-CY5
10372-932EA 776.19 USD
10372-932
Anti-PLCB3 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Phospholipase C is a key enzyme in phosphatidylinositol (PIP2) metabolism and lipid signaling pathways. The Phospholipase C family consists of 13 isozymes split between six subfamilies. Phospholipase C beta is activated by G protein subunits.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PLCB3
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR